PRI424048599 Princeton BioMolecular Research, Inc.

1-(2,4-dichlorobenzyl)-1H-pyrazol-4-amine

  • MDL number MFCD02055831
  • Cas Number : 514801-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart