PRI424048600 Princeton BioMolecular Research, Inc.

1-(4-bromobenzyl)-1H-pyrazol-4-amine

  • MDL number MFCD02253994
  • Cas Number : 1002651-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart