PRI424048626 Princeton BioMolecular Research, Inc.

1-(3-(1H-pyrrol-1-yl)phenyl)ethanamine

  • MDL number MFCD04967276
  • Cas Number : 1431970-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart