PRI424048682 Princeton BioMolecular Research, Inc.

1-((3-fluorophenoxy)methyl)-3,5-dimethyl-1H-pyrazol-4-amine

  • MDL number MFCD04967714
  • Cas Number : 1004192-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart