PRI424048684 Princeton BioMolecular Research, Inc.

1-((4-chloro-3-methylphenoxy)methyl)-3,5-dimethyl-1H-pyrazol-4-amine

  • MDL number MFCD04967692
  • Cas Number : 956394-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart