PRI424048732 Princeton BioMolecular Research, Inc.

4-(4-carboxyphenyl)pyridine-2-carboxylic acid

  • MDL number MFCD08690698
  • Cas Number : 914207-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart