PRI424048737 Princeton BioMolecular Research, Inc.

2,6-ditert-butylisonicotinic acid

  • MDL number MFCD06496458
  • Cas Number : 191478-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart