PRI424048760 Princeton BioMolecular Research, Inc.

5-nitroquinoline-6-carbaldehyde

  • MDL number MFCD07506536
  • Cas Number : 101327-85-9
  • Molecular Weight : 202.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart