PRI424048952 Princeton BioMolecular Research, Inc.

6-((methylsulfonyl)methyl)pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD09754190
  • Cas Number : 944058-91-7
  • Molecular Weight : 204.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-[(methylsulfonyl)methyl]-2,4(1H,3H)-pyrimidinedione
Inchi Code 1S/C6H8N2O4S/c1-13(11,12)3-4-2-5(9)8-6(10)7-4/h2H,3H2,1H3,(H2,7,8,9,10)
Inchi Key QDUCJSGZWMZJAT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart