PRI424048972 Princeton BioMolecular Research, Inc.

5-(4-Chloro-phenyl)-furan-2-carbonyl chloride

  • MDL number MFCD01860296
  • Cas Number : 57753-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart