PRI424048978 Princeton BioMolecular Research, Inc.

3-Isobutoxy-benzoyl chloride

  • MDL number MFCD03031490
  • Cas Number : 443290-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart