PRI424048990 Princeton BioMolecular Research, Inc.

4-Morpholin-4-yl-benzene-1,3-diamine

  • MDL number MFCD01918901
  • Cas Number : 37878-67-4
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart