PRI424049018 Princeton BioMolecular Research, Inc.

(2-(dimethylamino)-2-oxoethoxy)acetic acid

  • MDL number MFCD09880305
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(dimethylamino)-2-oxoethoxy]acetic acid
Inchi Code 1S/C6H11NO4/c1-7(2)5(8)3-11-4-6(9)10/h3-4H2,1-2H3,(H,9,10)
Inchi Key VYWKDPDSVACTBN-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart