PRI424049019 Princeton BioMolecular Research, Inc.

N-(3-aminopropyl)-1,5-dimethyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD09880306
  • Cas Number : 1174307-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart