PRI424049022 Princeton BioMolecular Research, Inc.

1-(1-(4-chlorophenyl)ethyl)-1H-pyrazol-5-amine

  • MDL number MFCD09738612
  • Cas Number : 1052547-97-3
  • Molecular Weight : 221.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[1-(4-chlorophenyl)ethyl]-1H-pyrazol-5-ylamine
Inchi Code 1S/C11H12ClN3/c1-8(15-11(13)6-7-14-15)9-2-4-10(12)5-3-9/h2-8H,13H2,1H3
Inchi Key RFIPVJJGXFMAKY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart