PRI424049026 Princeton BioMolecular Research, Inc.

2,5-diethylbenzenesulfonyl chloride

  • MDL number MFCD00625743
  • Cas Number : 19859-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart