PRI424049041 Princeton BioMolecular Research, Inc.

(2-(diethylamino)-2-oxoethoxy)acetic acid

  • MDL number MFCD06066356
  • Cas Number : 56074-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart