PRI424049057 Princeton BioMolecular Research, Inc.

2-(3,4-Dimethoxy-phenyl)-2-piperidin-1-yl-ethylamine

  • MDL number MFCD08445729
  • Cas Number : 889939-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart