PRI424049059 Princeton BioMolecular Research, Inc.

2-Piperidin-1-yl-thiazol-4-ylamine

  • MDL number MFCD09802061
  • Cas Number : 754954-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart