PRI424049084 Princeton BioMolecular Research, Inc.

4-tert-butyl-6-chloro-2-pyrrolidin-1-ylpyrimidine

  • MDL number MFCD09880755
  • Cas Number : 1023815-52-2
  • Molecular Weight : 239.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-tert-butyl-6-chloro-2-(1-pyrrolidinyl)pyrimidine
Inchi Code 1S/C12H18ClN3/c1-12(2,3)9-8-10(13)15-11(14-9)16-6-4-5-7-16/h8H,4-7H2,1-3H3
Inchi Key WZGKRRLOVIQNTN-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart