PRI424049085 Princeton BioMolecular Research, Inc.

4-(4-tert-butyl-6-chloropyrimidin-2-yl)morpholine

  • MDL number MFCD09880760
  • Cas Number : 1023815-62-4
  • Molecular Weight : 255.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-tert-butyl-6-chloro-2-pyrimidinyl)morpholine
Inchi Code 1S/C12H18ClN3O/c1-12(2,3)9-8-10(13)15-11(14-9)16-4-6-17-7-5-16/h8H,4-7H2,1-3H3
Inchi Key QEZYJPLTBFIOQS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart