PRI424049107 Princeton BioMolecular Research, Inc.

1-(4-fluorophenyl)-N~3~,N~3~-dimethylpropane-1,3-diamine

  • MDL number MFCD09880886
  • Cas Number : 933700-53-9
  • Molecular Weight : 196.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-fluorophenyl)-N~3~,N~3~-dimethyl-1,3-propanediamine
Inchi Code 1S/C11H17FN2/c1-14(2)8-7-11(13)9-3-5-10(12)6-4-9/h3-6,11H,7-8,13H2,1-2H3
Inchi Key JWUZFHYLKAZQPJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart