PRI424049163 Princeton BioMolecular Research, Inc.

1-(Biphenyl-4-carbonyl)-piperidine-4-carboxylic acid

  • MDL number MFCD04209768
  • Molecular Weight : 309.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-([1,1'-biphenyl]-4-ylcarbonyl)-4-piperidinecarboxylic acid
Inchi Code 1S/C19H19NO3/c21-18(20-12-10-17(11-13-20)19(22)23)16-8-6-15(7-9-16)14-4-2-1-3-5-14/h1-9,17H,10-13H2,(H,22,23)
Inchi Key GABASQJONDKFTJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart