PRI424049188 Princeton BioMolecular Research, Inc.

4-chloropteridine

  • MDL number MFCD08061287
  • Cas Number : 72700-48-2
  • Molecular Weight : 166.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart