PRI424049189 Princeton BioMolecular Research, Inc.

6-chloro-8-methyl-9H-purine

  • MDL number MFCD00234163
  • Cas Number : 92001-52-0
  • Molecular Weight : 168.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart