PRI424049197 Princeton BioMolecular Research, Inc.

3-bromo-4-hydroxyfuran-2(5H)-one

  • MDL number MFCD09863924
  • Cas Number : 21151-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart