PRI424049231 Princeton BioMolecular Research, Inc.

2-(((3,5-dichlorophenyl)amino)carbonyl)cyclohexanecarboxylic acid

  • MDL number MFCD01835015
  • Molecular Weight : 316.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart