PRI424049247 Princeton BioMolecular Research, Inc.

4,6-dimethylpyridine-2-carbaldehyde

  • MDL number MFCD00160403
  • Cas Number : 5439-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart