PRI424049343 Princeton BioMolecular Research, Inc.

diisopropyl 2-oxaspiro[3.3]heptane-6,6-dicarboxylate

  • MDL number MFCD09881398
  • Cas Number : 1023817-50-6
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diisopropyl 2-oxaspiro[3.3]heptane-6,6-dicarboxylate
Inchi Code 1S/C14H22O5/c1-9(2)18-11(15)14(12(16)19-10(3)4)5-13(6-14)7-17-8-13/h9-10H,5-8H2,1-4H3
Inchi Key SBZAGERPZYUHPH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart