PRI424049439 Princeton BioMolecular Research, Inc.

4,5-diamino-2-methylpyridazin-3(2H)-one

  • MDL number MFCD11106732
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart