PRI424049536 Princeton BioMolecular Research, Inc.

2-Phenyl-3H-pteridin-4-one

  • MDL number MFCD11107277
  • Cas Number : 23120-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart