PRI424049549 Princeton BioMolecular Research, Inc.

3-Bromo-2-pyridin-4-yl-1H-indole

  • MDL number MFCD02068317
  • Cas Number : 343350-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart