PRI424049570 Princeton BioMolecular Research, Inc.

4-Chloro-5-(2-chloro-ethyl)-6-methyl-pyrimidin-2-ylamine

  • MDL number MFCD11107442
  • Cas Number : 306314-90-9
  • Molecular Weight : 206.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-5-(2-chloroethyl)-6-methyl-2-pyrimidinylamine
Inchi Code 1S/C7H9Cl2N3/c1-4-5(2-3-8)6(9)12-7(10)11-4/h2-3H2,1H3,(H2,10,11,12)
Inchi Key WYERPOXPYAEELU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart