PRI424049576 Princeton BioMolecular Research, Inc.

6-Methoxy-benzo[1,2,4]triazine-3-carboxylic acid

  • MDL number MFCD11107470
  • Molecular Weight : 205.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methoxy-1,2,4-benzotriazine-3-carboxylic acid
Inchi Code 1S/C9H7N3O3/c1-15-5-2-3-6-7(4-5)10-8(9(13)14)12-11-6/h2-4H,1H3,(H,13,14)
Inchi Key YMTVNVHHYPCSLK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart