PRI424049680 Princeton BioMolecular Research, Inc.

4-chloro-6-methylnicotinamide

  • MDL number MFCD11108707
  • Cas Number : 473255-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart