PRI424049698 Princeton BioMolecular Research, Inc.

2-amino-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxamide

  • MDL number MFCD08694042
  • Cas Number : 24248-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart