PRI424049715 Princeton BioMolecular Research, Inc.

N-{[1-(tert-butoxycarbonyl)piperidin-4-yl]carbonyl}-beta-alanine

  • MDL number MFCD11109065
  • Molecular Weight : 300.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-{[1-(tert-butoxycarbonyl)-4-piperidinyl]carbonyl}-beta-alanine
Inchi Code 1S/C14H24N2O5/c1-14(2,3)21-13(20)16-8-5-10(6-9-16)12(19)15-7-4-11(17)18/h10H,4-9H2,1-3H3,(H,15,19)(H,17,18)
Inchi Key PCXAITGDJOJZGM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart