PRI424049721 Princeton BioMolecular Research, Inc.

C-[1-(4-Methyl-piperidin-1-yl)-cyclohexyl]-methylamine

  • MDL number MFCD08002285
  • Cas Number : 889939-96-2
  • Molecular Weight : 210.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(4-methyl-1-piperidinyl)cyclohexyl]methanamine
Inchi Code 1S/C13H26N2/c1-12-5-9-15(10-6-12)13(11-14)7-3-2-4-8-13/h12H,2-11,14H2,1H3
Inchi Key QBONHBWSWAFTTK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart