PRI424049733 Princeton BioMolecular Research, Inc.

2-(pyridin-4-ylamino)benzoic acid

  • MDL number MFCD11556238
  • Cas Number : 34861-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart