PRI424049843 Princeton BioMolecular Research, Inc.

(3-tert-butyl-1-methyl-1H-pyrazol-4-yl)methylamine

  • MDL number MFCD09965654
  • Cas Number : 1174309-25-1
  • Molecular Weight : 167.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart