PRI424049903 Princeton BioMolecular Research, Inc.

5-(2-furoylamino)-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11868694
  • Cas Number : 1174308-67-8
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart