PRI424049979 Princeton BioMolecular Research, Inc.

5,8-dichloroquinoline-2-carbaldehyde

  • MDL number MFCD06824166
  • Molecular Weight : 226.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart