PRI424050008 Princeton BioMolecular Research, Inc.

2,3-dimethyl-1H-indole-7-carbonitrile

  • MDL number MFCD12402553
  • Cas Number : 103503-07-7
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-dimethyl-1H-indole-7-carbonitrile
Inchi Code 1S/C11H10N2/c1-7-8(2)13-11-9(6-12)4-3-5-10(7)11/h3-5,13H,1-2H3
Inchi Key DECFHWWJJPYUKQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart