PRI424050014 Princeton BioMolecular Research, Inc.

(4'-chloro-1,1'-biphenyl-2-yl)methylamine

  • MDL number MFCD05981938
  • Cas Number : 153850-88-5
  • Molecular Weight : 217.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart