PRI424050017 Princeton BioMolecular Research, Inc.

2'-fluoro-1,1'-biphenyl-3-carbaldehyde

  • MDL number MFCD03424637
  • Cas Number : 676348-33-7
  • Molecular Weight : 200.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart