PRI424050029 Princeton BioMolecular Research, Inc.

(6-piperazin-1-ylpyridin-3-yl)methanol

  • MDL number MFCD12403471
  • Cas Number : 1216164-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart