PRI424050050 Princeton BioMolecular Research, Inc.

methyl 4-(aminomethyl)-5-methyl-2-furoate

  • MDL number MFCD12403732
  • Cas Number : 748707-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart