PRI424050151 Princeton BioMolecular Research, Inc.

2-(2-Methyl-5-trifluoromethoxy-1H-indol-3-yl)-ethylamine

  • MDL number MFCD02656518
  • Molecular Weight : 258.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart