PRI424050247 Princeton BioMolecular Research, Inc.

3-Chloro-5-methoxy-benzo[b]thiophene-2-carbonyl chloride

  • MDL number MFCD09965821
  • Cas Number : 56030-16-1
  • Molecular Weight : 261.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-5-methoxy-1-benzothiophene-2-carbonyl chloride
Inchi Code 1S/C10H6Cl2O2S/c1-14-5-2-3-7-6(4-5)8(11)9(15-7)10(12)13/h2-4H,1H3
Inchi Key DLJHFQNHWUTVRD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart