PRI424050257 Princeton BioMolecular Research, Inc.

3-Bromo-5-methyl-1H-indazole

  • MDL number MFCD07781622
  • Cas Number : 40598-72-9
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart